Learn More
Medchemexpress LLC HY-130243 5mg Medchemexpress, RORγt Inverse agonist 6 CAS:1887161-80-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1302435MG
Encompass_Preferred
Medchemexpress, HY-130243 5mg RORγt Inverse agonist 6 CAS:1887161-80-9 RORγt Inverse agonist 6 (compound 43) is a RORγt inverse agonist for the study of Th17-driven autoimmune diseases. RORγt Inverse agonist 6 (compound 43) suppresses IL-17A gene expression by IL-23 stimulation in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1231