Learn More
Eurofins Discoverx Ros, active, 10 µg. (14-527)
Supplier:  Eurofins Discoverx 14527
Encompass
N-terminal 6xHis-tagged recombinant, human Ros amino acids 1883-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8923