Learn More
Cayman Chemical ROS Detection Cell-Based Assay Kit (DCFDA), 96 Well
SDP
Catalog No. 502893767
Encompass_Preferred
Cayman’s ROS Detection Cell-Based Assay Kit (DCFDA) uses DCFDA as a fluorescent probe for the detection of ROS generation. Pyocyanin, a cell-permeable compound capable of redox cycling, is included as a positive control for ROS generation. N-acetyl-cysteine is included as an antioxidant control.
Catalog No. 50-289-3767 Supplier Cayman Chemical Supplier No. 60152096 WELL
Cayman’s ROS Detection Cell-Based Assay Kit (DCFDA) uses DCFDA as a fluorescent probe for the detection of ROS generation. Pyocyanin, a cell-permeable compound capable of redox cycling, is included as a positive control for ROS generation. N-acetyl-cysteine is included as an antioxidant control.