Learn More
Cell Signaling Technology Monoclonal Antibody, ROS1, Mouse, Human Reactivity, 100 μg
SDP
Catalog No. 503132998
Encompass
This antibody recognizes endogenous levels of total ROS1 protein without cross-reacting with other related proteins. Produced by immunizing animals with a GST-ROS1 fusion protein specific to the carboxy terminus of human ROS1, this monoclonal antibody is provided in a BSA and azide-free formulation. It is ideal for specialized or custom antibody labeling applications, such as the conjugation of fluorophores, metals, lanthanides, or oligonucleotides.
  • Validated for Western blotting and ELISA-based applications
  • Specifically recognizes human ROS1
  • Carrier-free formulation suitable for custom labeling
  • Quality controlled via size exclusion chromatography to ensure antibody integrity
Catalog No. 50-313-2998 Supplier Cell Signaling Technology Supplier No. 35744SF
This antibody recognizes endogenous levels of total ROS1 protein without cross-reacting with other related proteins. Produced by immunizing animals with a GST-ROS1 fusion protein specific to the carboxy terminus of human ROS1, this monoclonal antibody is provided in a BSA and azide-free formulation. It is ideal for specialized or custom antibody labeling applications, such as the conjugation of fluorophores, metals, lanthanides, or oligonucleotides.
  • Validated for Western blotting and ELISA-based applications
  • Specifically recognizes human ROS1
  • Carrier-free formulation suitable for custom labeling
  • Quality controlled via size exclusion chromatography to ensure antibody integrity