Learn More
Amsbio LLC ROS1 positive non-small cell l
Catalog No. 502564346
Encompass
ROS1 positive non-small cell lung NSCLC cancer tissue array including pathology grade TNM clinical stage and IHC ROS1+ results 21 cases/26 cores
Catalog No. 50-256-4346 Supplier Amsbio LLC Supplier No. LC261
ROS1 positive non-small cell lung NSCLC cancer tissue array including pathology grade TNM clinical stage and IHC ROS1+ results 21 cases/26 cores