Learn More
Cayman Chemical OM173-αA, Purity: ≥95%, CAS: 58286-56-9, 5 mg
SDP
Catalog No. 502940939
Encompass_Preferred
A quinone bacterial metabolite; inhibits the growth of the bacteria M. gallisepticum, M. pneumoniae, and S. aureus in vitro (MICs = 0.39-3.13 μg/ml); inhibits the growth of the plant pathogenic fungus P. oryzae (MIC = 3.12 μg/ml) and several species of Trichophyton (MICs = 12.5-25 μg/ml)
Catalog No. 50-294-0939 Supplier Cayman Chemical Supplier No. 275705 MG
A quinone bacterial metabolite; inhibits the growth of the bacteria M. gallisepticum, M. pneumoniae, and S. aureus in vitro (MICs = 0.39-3.13 μg/ml); inhibits the growth of the plant pathogenic fungus P. oryzae (MIC = 3.12 μg/ml) and several species of Trichophyton (MICs = 12.5-25 μg/ml)