Learn More
Cayman Chemical Rosamultin, Purity: ≥98%, CAS: 88515-58-6, 10 mg
SDP
Catalog No. 502945921
Encompass_Preferred
A triterpenoid glycoside with diverse biological activities; inhibits HIV-1 protease by 52.9% in a cell-free assay at 100 µM; reduces hydrogen peroxide-induced apoptosis, increases in MDA levels, and decreases in Sod, Cat, and Gpx activities in H9c2 rat cardiomyocytes at 0.01 nM; increases survival, prevents decreases in body weight, and protects against splenic and intestinal damage in irradiated mice at 12 mg/kg; inhibits acetic acid-induced writhing in mice at 30 mg/kg, p.o.
Catalog No. 50-294-5921 Supplier Cayman Chemical Supplier No. 3993910 MG
A triterpenoid glycoside with diverse biological activities; inhibits HIV-1 protease by 52.9% in a cell-free assay at 100 µM; reduces hydrogen peroxide-induced apoptosis, increases in MDA levels, and decreases in Sod, Cat, and Gpx activities in H9c2 rat cardiomyocytes at 0.01 nM; increases survival, prevents decreases in body weight, and protects against splenic and intestinal damage in irradiated mice at 12 mg/kg; inhibits acetic acid-induced writhing in mice at 30 mg/kg, p.o.