Learn More
Selleck Chemical LLC Roscovitine (Seliciclib) 50mg 186692-46-6 CYC202
SDP
Catalog No. 508362
Encompass_Preferred
Roscovitine (Seliciclib, CYC202) is a potent and selective CDK inhibitor for Cdc2, CDK2 and CDK5 with IC50 of 0.65 ?M, 0.7 ?M and 0.16 ?M in cell-free assays. It shows little effect on CDK4/6. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-836-2 Supplier Selleck Chemical LLC Supplier No. S115350MG
Roscovitine (Seliciclib, CYC202) is a potent and selective CDK inhibitor for Cdc2, CDK2 and CDK5 with IC50 of 0.65 ?M, 0.7 ?M and 0.16 ?M in cell-free assays. It shows little effect on CDK4/6. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.