Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Rose Agar (CNA and PEA)
SDP

Catalog No. 23001739 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-739 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-739 Supplier Hardy Diagnostics™ Supplier No. A66
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rose agar is recommended for use as a selective growth medium for the cultivation and isolation of gram-positive cocci from clinical and non-clinical specimens which contain mixed flora.

  • For the selective cultivation and isolation of gram-positive cocci.
  • Rose agar consists of half Columbia CNA Agar and Half Phenylethyl Alcohol Agar Base
  • Contains 5% sheep blood from a veterinary supervised, controlled herd of donor animals. Blood is guaranteed to be from healthy antibiotic-free animals.
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding.
  • Packaged in a sleeve of 10 in a celluose bag that prevents build-up of condensation and excess moisture.

Specifications

Color Red
Content And Storage 2°C to 8°C
Additive Phenylethanol, Colistin Sulfate, Nalidixic Acid, Sheep Blood
Detects Gram-positive Cocci
Format Plate
Media Type Rose Agar
Product Type Agar Plate
Sterility Sterile
Target Organism Class Gram-positive Cocci
Test Type Microbial Cultivation
Fill Volume 17 mL
Formulation Pancreatic and Enzymatic Digests of Casein - 25.0gm, Casein Yeast Peptone - 10.0gm, Sodium Chloride - 10.0gm, Papaic Digest of Soybean Meal - 5.0gm, Tryptic Digest of Beef Heart - 3.0gm, Phenylethanol - 2.5gm, Corn Starch - 1.0gm, Colistin Sulfate - 8.25mg, Nalidixic Acid - 4.0mg, Sheep Blood - 50.0ml, Agar - 30.0gm
For Use With (Application) Cultivation and isolation of gram-positive cocci
Incubation Conditions 35°C to 37°C, Aerobic
Incubation Time 24 hr.
Material Polystyrene
No. per Pack 10
Packaging Type Sleeve, Breathable Bag
Quantity 10 Plates
Dimensions (Dia. x H) 100 x 15 mm
Width (Metric) 100 mm
Height (English) 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Streptococcus species, Proteus mirabilis, Pseudomonas aeruginosa
Protein Source Casein, Yeast, Soybean, Beef Heart, Sheep Blood
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.