Learn More
Medchemexpress LLC Rose bengal | 11121-48-5 | MFCD00005043 | 95.0% | 1017.6 g/mol | C20H2Cl4I4Na2O5 | 5 G
SDP
Catalog No. 5000281967
Encompass_Preferred
Rose bengal is a xanthene dye and photosensitizer intended for research use. It functions as a staining agent and photodynamic antimicrobial agent and is used in biological staining, photochemical studies, and analytical applications. The material is supplied as a solid powder with recommended sealed storage away from light and moisture.
  • Photosensitizer dye for photodynamic studies.
  • Used as a staining agent and analytical dye.
  • Applicable to antimicrobial and photochemical research.
  • Supplied as a solid powder with sealed storage recommendations.
  • CAS number 11121-48-5 for unambiguous identification.
Catalog No. 50-002-81967 Supplier Medchemexpress LLC Supplier No. HYW3290715G
Rose bengal is a xanthene dye and photosensitizer intended for research use. It functions as a staining agent and photodynamic antimicrobial agent and is used in biological staining, photochemical studies, and analytical applications. The material is supplied as a solid powder with recommended sealed storage away from light and moisture.
  • Photosensitizer dye for photodynamic studies.
  • Used as a staining agent and analytical dye.
  • Applicable to antimicrobial and photochemical research.
  • Supplied as a solid powder with sealed storage recommendations.
  • CAS number 11121-48-5 for unambiguous identification.