Learn More
MOLTOX Molecular Toxicology Inc Rose Bengal Agar, Contact Plate, 10 plates/sleeve, Store in Refrigerator
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 2140S68
Encompass_Preferred
Rose Bengal Agar is used in conjunction with chloramphenicol for isolating and enumerating yeasts and molds from food and water samples. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-683