Learn More
Description
Rose bengal has several microbilogical staining applications including: Conn's technique for bacteria in soil and Kreyberg's stain for keratin and mucuscounterstaining hematoxylinstaining cellular inclusions, together with Bismark brown
Specifications
Specifications
| CAS | 632-69-9 |
| Concentration | ≥80 % |
| Quantity | 25 g |
| Product Type | Stain Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.