Learn More
Abcam Rose bengal diacetate, 5MG
Supplier:  Abcam AB2754915MG
Encompass_Preferred
MW 1057.7 Da, Purity >98%. Diacetate derivative of rose bengal with greatly improved membrane permeability, allowing it to accumulate intracellularly where esterase activity releases the active rose bengal photosensitizer. Activation with UV light at 561 nm causes singlet oxygen production and photosensitization.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-134