Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rose Bengal, MP Biomedicals™

Catalog No. ICN19549105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19549105 5 g
ICN19549125 25 g
ICN19549180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19549105 Supplier MP Biomedicals, Inc Supplier No. 0219549105
Add to Cart
Add to Cart

Chemical Identifiers

CAS 632-68-8
Molecular Formula C20H2Cl4I4K2O5
Molecular Weight (g/mol) 1049.845
MDL Number MFCD00005043
InChI Key VQHHOXOLUXRQFQ-UHFFFAOYSA-L
Synonym Acid Red 94, C.I. 45440
PubChem CID 71553
IUPAC Name dipotassium;4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate
SMILES C1=C2C(=C(C(=C1I)[O-])I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)[O-])I.[K+].[K+]

Specifications

Formula Weight 1017.65
Color Red to Rust
Quantity 5 g
Physical Form Powder
Chemical Name or Material Rose Bengal
EINECSNumber 211-183-3
Recommended Storage Store at room temperature.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.