Learn More
TARGETMOL CHEMICALS INC ROSE BENGAL SODIUM 500MG
SDP
Catalog No. 502579165
Encompass_Preferred
Also available in 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rose Bengal sodium is a potent inhibitor of VGlut and vesicular monoamine transporter (VMAT) (Ki of 19 and 64 nM for VGlut andVMAT respectively)purity: 91%
Catalog No. 50-257-9165 Supplier TARGETMOL CHEMICALS INC Supplier No. T7651500MG
Also available in 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rose Bengal sodium is a potent inhibitor of VGlut and vesicular monoamine transporter (VMAT) (Ki of 19 and 64 nM for VGlut andVMAT respectively)purity: 91%