Learn More
Cayman Chemical Rose BengalsodIum salt 50g
SDP
Catalog No. 502939063
Encompass_Preferred
A xanthene dye, fluorescein derivative, and photosensitizer; abs/em maxima = 548/567 nm, respectively; binds to S. aureus cells and decreases survival of photoirradiated S. aureus to 0.012% at 1 μM; generates singlet oxygen when exposed to photoirradiation in cell-free assays; induces potassium ion leakage from S. aureus and bovine erythrocytes in the presence of photoirradiation; inhibits CYP3A4/5 and UGT1A6 in human liver microsomes in a light-dependent manner (IC50s = 0.072 and 0.035 μM, respectively, in yellow light, 3.1 and 4.2 μM, respectively, in ambient light, and 3 and 4.2 μM, respectively, in the dark); has been used for staining of live cells, but exhibits both intrinsic and phototoxicity
Catalog No. 50-293-9063 Supplier Cayman Chemical Supplier No. 2882950 G
A xanthene dye, fluorescein derivative, and photosensitizer; abs/em maxima = 548/567 nm, respectively; binds to S. aureus cells and decreases survival of photoirradiated S. aureus to 0.012% at 1 μM; generates singlet oxygen when exposed to photoirradiation in cell-free assays; induces potassium ion leakage from S. aureus and bovine erythrocytes in the presence of photoirradiation; inhibits CYP3A4/5 and UGT1A6 in human liver microsomes in a light-dependent manner (IC50s = 0.072 and 0.035 μM, respectively, in yellow light, 3.1 and 4.2 μM, respectively, in ambient light, and 3 and 4.2 μM, respectively, in the dark); has been used for staining of live cells, but exhibits both intrinsic and phototoxicity