Learn More
Medchemexpress LLC Rose-β-D-gal | 138182-21-5 | MFCD00467206 | 99.7% | 329.73 g/mol | C14H16ClNO6 | 25 MG
SDP
Catalog No. 5000282589
Encompass_Preferred
Rose-β-D-Gal is a chromogenic β-galactosidase substrate that produces a pink/magenta color upon enzymatic cleavage. It is used to detect β-gal activity in biochemical and cell-based assays where a visible color change is required.
  • Produces a pink/magenta chromogenic signal for β-gal activity detection.
  • Suitable for enzymatic assays and staining applications.
  • High reported purity (99.71%) for consistent performance.
  • Solid reagent supplied in small milligram quantities for research use.
  • Compatible with colorimetric readouts and microscopy.
Catalog No. 50-002-82589 Supplier Medchemexpress LLC Supplier No. HYW12776925MG
Rose-β-D-Gal is a chromogenic β-galactosidase substrate that produces a pink/magenta color upon enzymatic cleavage. It is used to detect β-gal activity in biochemical and cell-based assays where a visible color change is required.
  • Produces a pink/magenta chromogenic signal for β-gal activity detection.
  • Suitable for enzymatic assays and staining applications.
  • High reported purity (99.71%) for consistent performance.
  • Solid reagent supplied in small milligram quantities for research use.
  • Compatible with colorimetric readouts and microscopy.