Learn More
Medchemexpress LLC Rose-β-D-Gal | 138182-21-5 | MFCD01310928 | 99.7% | 329.73 g/mol | C14H16ClNO6 | 10 MG
SDP
Catalog No. 5000282588
Encompass_Preferred
Rose-β-D-Gal is a chromogenic substrate for β-galactosidase that produces a pink/magenta color upon enzymatic cleavage. It exhibits fluorescent properties (excitation 488 nm and emission 540 nm), is supplied as a white to off-white solid, and is intended for biochemical and cell-based assays to detect β-gal activity.
  • Chromogenic substrate for β-galactosidase detection.
  • Produces a pink/magenta color upon enzymatic cleavage.
  • Excitation 488 nm and emission 540 nm for fluorescence-based assays.
  • High purity suitable for analytical and research applications.
  • Solid form for easy weighing and storage.
Catalog No. 50-002-82588 Supplier Medchemexpress LLC Supplier No. HYW12776910MG
Rose-β-D-Gal is a chromogenic substrate for β-galactosidase that produces a pink/magenta color upon enzymatic cleavage. It exhibits fluorescent properties (excitation 488 nm and emission 540 nm), is supplied as a white to off-white solid, and is intended for biochemical and cell-based assays to detect β-gal activity.
  • Chromogenic substrate for β-galactosidase detection.
  • Produces a pink/magenta color upon enzymatic cleavage.
  • Excitation 488 nm and emission 540 nm for fluorescence-based assays.
  • High purity suitable for analytical and research applications.
  • Solid form for easy weighing and storage.