Learn More
Bioworld Rose Initiation Medium, 25 L
Catalog No. 50254929
Encompass_Preferred
Rose Initiation MediumStage IContains Micronutrients and Macronutrients required for rose initiationContains FeNaEDTA in place of Ferrous Sulfate and Disodium EDTASucrose, and additional supplement should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.51 g/LpH: 3.0–4.0Soluble in water
Catalog No. 50-254-929 Supplier Bioworld Supplier No. 306301733
Rose Initiation MediumStage IContains Micronutrients and Macronutrients required for rose initiationContains FeNaEDTA in place of Ferrous Sulfate and Disodium EDTASucrose, and additional supplement should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.51 g/LpH: 3.0–4.0Soluble in water