Learn More
Bioworld Rose Initiation Medium, 50 L
Catalog No. 50254930
Encompass_Preferred
Rose Initiation MediumStage IContains Micronutrients and Macronutrients required for rose initiationContains FeNaEDTA in place of Ferrous Sulfate and Disodium EDTASucrose, and additional supplement should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.51 g/LpH: 3.0–4.0Soluble in water
Catalog No. 50-254-930 Supplier Bioworld Supplier No. 306301734
Rose Initiation MediumStage IContains Micronutrients and Macronutrients required for rose initiationContains FeNaEDTA in place of Ferrous Sulfate and Disodium EDTASucrose, and additional supplement should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.51 g/LpH: 3.0–4.0Soluble in water