Learn More
Cayman Chemical RoseoflavIn 10mg
SDP
Catalog No. 502972947
Encompass_Preferred
A riboflavin analog that possesses antimicrobial activity; binds directly to FMN riboswitch aptamers (KD = ~100 nM), downregulating the expression of genes responsible for the synthesis and transport of riboflavin
Catalog No. 50-297-2947 Supplier Cayman Chemical Supplier No. 1600110 MG
A riboflavin analog that possesses antimicrobial activity; binds directly to FMN riboswitch aptamers (KD = ~100 nM), downregulating the expression of genes responsible for the synthesis and transport of riboflavin