Learn More
STEMCELL Technologies RosetteSep™ Human CD4+ T Cell Enrichment Cocktail, For processing: 40 mL whole blood processed
SDP
Catalog No. NC0237030
Encompass
The RosetteSep™ Human CD4+ T Cell Enrichment Cocktail is designed to isolate CD4+ T cells from whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-CD4+ T cells and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs. The purified CD4+ T cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium.
Catalog No. NC0237030 Supplier STEMCELL Technologies Supplier No. 15022
The RosetteSep™ Human CD4+ T Cell Enrichment Cocktail is designed to isolate CD4+ T cells from whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-CD4+ T cells and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs. The purified CD4+ T cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium.