Learn More
STEMCELL Technologies RosetteSep CTC Enrichment Cocktail Containing Anti-CD56, For processing: 200 mL whole blood processed
SDP
Catalog No. 502390101
Encompass
The RosetteSep CTC Enrichment Cocktail Containing Anti-CD56 is designed to enrich circulating epithelial tumor cells from fresh whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD3 CD14 CD16 CD19 CD38 CD45 CD56 CD61 CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a density gradient medium such as Lymphoprep (Catalog 07801) the unwanted cells pellet along with the RBCs. The purified tumor cells are present as a highly enriched population at the interface between the plasma and the density gradient medium.
Catalog No. 50-239-0101 Supplier STEMCELL Technologies Supplier No. 15177
The RosetteSep CTC Enrichment Cocktail Containing Anti-CD56 is designed to enrich circulating epithelial tumor cells from fresh whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD3 CD14 CD16 CD19 CD38 CD45 CD56 CD61 CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a density gradient medium such as Lymphoprep (Catalog 07801) the unwanted cells pellet along with the RBCs. The purified tumor cells are present as a highly enriched population at the interface between the plasma and the density gradient medium.