Learn More
STEMCELL Technologies RosetteSep™ DM-M Density Medium, Size: 100 mL
SDP
Catalog No. 501974536
Encompass
RosetteSep™ DM-M is a density gradient medium designed specifically for use with RosetteSep™ cocktails for the enrichment of specific human myeloid (CD33+) cells from whole blood. RosetteSep™ DM-M has a density of 1.085 g/mL.Please Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled “For Research Use Only”. Please note there have been no changes to the form, function and quality assurances associated with these products. For more information please contact the Quality Assurance and Regulatory Department at qaschangenote@stemcell.com.
Catalog No. 50-197-4536 Supplier STEMCELL Technologies Supplier No. 15725
RosetteSep™ DM-M is a density gradient medium designed specifically for use with RosetteSep™ cocktails for the enrichment of specific human myeloid (CD33+) cells from whole blood. RosetteSep™ DM-M has a density of 1.085 g/mL.Please Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled “For Research Use Only”. Please note there have been no changes to the form, function and quality assurances associated with these products. For more information please contact the Quality Assurance and Regulatory Department at qaschangenote@stemcell.com.