Learn More
STEMCELL Technologies RosetteSep™ Human Bone Marrow Progenitor Cell Pre-Enrichment Cocktail, For processing: 40 mL bone marrow processed
SDP
Catalog No. 501974500
Encompass
The RosetteSep™ Human Bone Marrow Progenitor Cell Pre-Enrichment Cocktail is designed to pre-enrich progenitor cells by removing lineage-specific cells from whole bone marrow by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD3, CD11b, CD14, CD16, CD19, CD56, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs. The pre-enriched progenitor cells are present as an enriched population at the interface between the plasma and the buoyant density medium.
Catalog No. 50-197-4500 Supplier STEMCELL Technologies Supplier No. 15027
The RosetteSep™ Human Bone Marrow Progenitor Cell Pre-Enrichment Cocktail is designed to pre-enrich progenitor cells by removing lineage-specific cells from whole bone marrow by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD3, CD11b, CD14, CD16, CD19, CD56, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs. The pre-enriched progenitor cells are present as an enriched population at the interface between the plasma and the buoyant density medium.