Learn More
STEMCELL Technologies RosetteSep Human CD8 Depletion Cocktail, For processing: 200 mL whole blood processed
SDP
Catalog No. 502390386
Encompass
The RosetteSep Human CD8 Depletion Cocktail is designed to deplete CD8 cells from whole blood. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD8 and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep (Catalog 07801) the unwanted cells pellet along with the RBCs. The CD8 depleted fraction is present as a highly enriched population at the interface between the plasma and the buoyant density medium.
Catalog No. 50-239-0386 Supplier STEMCELL Technologies Supplier No. 15663
The RosetteSep Human CD8 Depletion Cocktail is designed to deplete CD8 cells from whole blood. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD8 and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep (Catalog 07801) the unwanted cells pellet along with the RBCs. The CD8 depleted fraction is present as a highly enriched population at the interface between the plasma and the buoyant density medium.