Learn More
STEMCELL Technologies RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktail, For processing: 40 mL cord blood processed
SDP
Catalog No. 501974499
Encompass
The RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktail is designed to isolate progenitor cells from cord blood and whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD3, CD14, CD16, CD19, CD24, CD56, CD61, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs. The purified progenitor cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium. If using large volumes of blood, we recommend the RosetteSep™ Cord Blood Progenitor Enrichment Kit with HetaSep™ (Catalog #15276) (/products/rosettesep-human-cord-blood-progenitor-enrichment-kit.html).
Catalog No. 50-197-4499 Supplier STEMCELL Technologies Supplier No. 15026
The RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktail is designed to isolate progenitor cells from cord blood and whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD3, CD14, CD16, CD19, CD24, CD56, CD61, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs. The purified progenitor cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium. If using large volumes of blood, we recommend the RosetteSep™ Cord Blood Progenitor Enrichment Kit with HetaSep™ (Catalog #15276) (/products/rosettesep-human-cord-blood-progenitor-enrichment-kit.html).