Learn More
STEMCELL Technologies RosetteSep Human Monocyte Depletion Cocktail, For processing: 200 mL whole blood processed
SDP
Catalog No. 502390188
Encompass
The RosetteSep Human Monocyte Depletion Cocktail is designed to deplete monocytes from whole blood. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD36 and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep (Catalog 07801) the unwanted cells pellet along with the RBCs. The monocyte depleted fraction is present as a highly enriched population at the interface between the plasma and the buoyant density medium.
Catalog No. 50-239-0188 Supplier STEMCELL Technologies Supplier No. 15668
The RosetteSep Human Monocyte Depletion Cocktail is designed to deplete monocytes from whole blood. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD36 and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep (Catalog 07801) the unwanted cells pellet along with the RBCs. The monocyte depleted fraction is present as a highly enriched population at the interface between the plasma and the buoyant density medium.