Learn More
STEMCELL Technologies RosetteSep™ Human Progenitor Cell Basic Pre-Enrichment Cocktail, For processing: 5000 mL cord blood processed
SDP
Catalog No. 501974523
Encompass
The RosetteSep™ Human Progenitor Cell Basic Pre-Enrichment Kit is designed to pre-enrich hematopoietic progenitor cells from cord blood by negative selection. Unwanted cells are targeted for removal with antibody complexes recognizing CD3, CD11b, CD19 and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs. The hematopoietic progenitor cells are present as an enriched population at the interface between the plasma and the buoyant density medium. Cells of interest can then be positively selected from the pre-enriched cell population using EasySep™ (e.g. EasySep™ Human CD34 Positive Selection).
Catalog No. 50-197-4523 Supplier STEMCELL Technologies Supplier No. 15266
The RosetteSep™ Human Progenitor Cell Basic Pre-Enrichment Kit is designed to pre-enrich hematopoietic progenitor cells from cord blood by negative selection. Unwanted cells are targeted for removal with antibody complexes recognizing CD3, CD11b, CD19 and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs. The hematopoietic progenitor cells are present as an enriched population at the interface between the plasma and the buoyant density medium. Cells of interest can then be positively selected from the pre-enriched cell population using EasySep™ (e.g. EasySep™ Human CD34 Positive Selection).