Learn More
Cayman Chemical Rosiglitazone-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1132641-22-5, 1 mg
SDP
Catalog No. 502986273
Encompass_Preferred
An internal standard for the quantification of rosiglitazone by GC- or LC-MS
Catalog No. 50-298-6273 Supplier Cayman Chemical Supplier No. 225621 MG
An internal standard for the quantification of rosiglitazone by GC- or LC-MS