Learn More
Enzo Life Sciences Rosiglitazone (100mg). CAS: 122320-73-4
SDP
Supplier:  Enzo Life Sciences ALX350125M100
Encompass_Preferred
Potent insulin sensitizing agent. Peroxisome proliferator-activated receptor γ (PPAR-γ) agonist. Hepatotoxic. Antidiabetic. Purity: ≥97% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (>25mg/ml), 100% ethanol or methanol. Long Term Storage: +4°C.
Catalog No. 50-200-8744