Learn More
Apexbio Technology LLC Rosiglitazone(Synonyms: Avandia, BRL-49653, Rosiglitazone Maleate, Rosiglitazone Base, BRL49653C, RGZ, Rosiglizole), 10mM (in 1mL DMSO), CAS: 122320-73-4.
SDP
Catalog No. 5000060532
Encompass_Preferred
Rosiglitazone (CAS 122320-73-4) is a synthetic thiazolidinedione (TZD) that functions as an agonist of peroxisome proliferator-activated receptor gamma (PPAR ) a nuclear receptor highly expressed in adipose tissues Upon ligand binding PPAR forms heterodimers with retinoid X receptors promoting transcriptional activation of genes regulating adipogenesis glucose uptake lipid metabolism and insulin sensitivity Through modulating fatty acid storage in adipose tissue and secretion of adipokines rosiglitazone significantly enhances insulin sensitivity making it extensively utilized in metabolic and diabetes research particularly type II diabetes studies
Catalog No. 50-000-60532 Supplier Apexbio Technology LLC Supplier No. A43045.1
Rosiglitazone (CAS 122320-73-4) is a synthetic thiazolidinedione (TZD) that functions as an agonist of peroxisome proliferator-activated receptor gamma (PPAR ) a nuclear receptor highly expressed in adipose tissues Upon ligand binding PPAR forms heterodimers with retinoid X receptors promoting transcriptional activation of genes regulating adipogenesis glucose uptake lipid metabolism and insulin sensitivity Through modulating fatty acid storage in adipose tissue and secretion of adipokines rosiglitazone significantly enhances insulin sensitivity making it extensively utilized in metabolic and diabetes research particularly type II diabetes studies