Learn More
Selleck Chemical LLC Rosiglitazone 10mM/1mL 122320-73-4 BRL 49653
SDP
Supplier:  Selleck Chemical LLC S255610MM/1ML
Encompass_Preferred
Rosiglitazone (BRL 49653) is a potent antihyperglycemic agent and a potent thiazolidinedione insulin sensitizer with IC50 of 12, 4 and 9 nM for rat, 3T3-L1 and human adipocytes, respectively. Rosiglitazone is a pure ligand of PPAR-gamma, and has no PPAR-alpha-binding action. Rosiglitazone modulates TRP channels and induces autophagy. Rosiglitazone prevents ferroptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2554