Learn More
Apexbio Technology LLC Rosiglitazone(Synonyms: Avandia, BRL-49653, Rosiglitazone Maleate, Rosiglitazone Base, BRL49653C, RGZ, Rosiglizole), 200mg, CAS: 122320-73-4.
SDP
Catalog No. 5000060802
Encompass_Preferred
Rosiglitazone (CAS 122320-73-4) is a synthetic thiazolidinedione (TZD) that functions as an agonist of peroxisome proliferator-activated receptor gamma (PPAR ) a nuclear receptor highly expressed in adipose tissues Upon ligand binding PPAR forms heterodimers with retinoid X receptors promoting transcriptional activation of genes regulating adipogenesis glucose uptake lipid metabolism and insulin sensitivity Through modulating fatty acid storage in adipose tissue and secretion of adipokines rosiglitazone significantly enhances insulin sensitivity making it extensively utilized in metabolic and diabetes research particularly type II diabetes studies
Catalog No. 50-000-60802 Supplier Apexbio Technology LLC Supplier No. A4304200
Rosiglitazone (CAS 122320-73-4) is a synthetic thiazolidinedione (TZD) that functions as an agonist of peroxisome proliferator-activated receptor gamma (PPAR ) a nuclear receptor highly expressed in adipose tissues Upon ligand binding PPAR forms heterodimers with retinoid X receptors promoting transcriptional activation of genes regulating adipogenesis glucose uptake lipid metabolism and insulin sensitivity Through modulating fatty acid storage in adipose tissue and secretion of adipokines rosiglitazone significantly enhances insulin sensitivity making it extensively utilized in metabolic and diabetes research particularly type II diabetes studies