Learn More
TARGETMOL CHEMICALS INC Rosiglitazone 200MG
SDP
Catalog No. 502880516
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Rosiglitazone (BRL49653) is a PPARgamma agonist, TRPC5 activator, and TRPM3 inhibitor with oral activity. Rosiglitazone is also a hypoglycemic agent and a thiazolidinedione insulin sensitizer. Purity 99.87%
Catalog No. 50-288-0516 Supplier TARGETMOL CHEMICALS INC Supplier No. T0334200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Rosiglitazone (BRL49653) is a PPARgamma agonist, TRPC5 activator, and TRPM3 inhibitor with oral activity. Rosiglitazone is also a hypoglycemic agent and a thiazolidinedione insulin sensitizer. Purity 99.87%