Learn More
AdipoGen Rosiglitazone,Chemical. CAS: 122320-73-4
SDP
Catalog No. 50464764
Encompass_Preferred
Rosiglitazone, AdipoGen Life Sciences. Chemical. CAS: 122320-73-4. Formula: C18H19N3O3S. MW: 357.4. Antidiabetic, hypoglycemic agent. Potent and selective peroxisome proliferator-activated receptor gamma (PPAR-gamma) agonist. Potent insulin sensitizing agent binding to the PPAR receptors in fat cells and making the cells more responsive to insulin. Ameliorates insulin resistance. Improves blood pressure and vascular function. Enhances proliferation of endogenous neural progenitor cells (NPCs). Anti-inflammatory compound. Has controversial therapeutic effects on the cardiovascular system. Promotes adipocyte differentiation of mesenchymal stem cells (MSCs).
Catalog No. 50-464-764 Supplier AdipoGen Supplier No. AGCR13570M025
Rosiglitazone, AdipoGen Life Sciences. Chemical. CAS: 122320-73-4. Formula: C18H19N3O3S. MW: 357.4. Antidiabetic, hypoglycemic agent. Potent and selective peroxisome proliferator-activated receptor gamma (PPAR-gamma) agonist. Potent insulin sensitizing agent binding to the PPAR receptors in fat cells and making the cells more responsive to insulin. Ameliorates insulin resistance. Improves blood pressure and vascular function. Enhances proliferation of endogenous neural progenitor cells (NPCs). Anti-inflammatory compound. Has controversial therapeutic effects on the cardiovascular system. Promotes adipocyte differentiation of mesenchymal stem cells (MSCs).