Learn More
Cayman Chemical Rosiglitazone, 122320-73-4, 25 mg
SDP
Supplier:  Cayman Chemical 7174025
Encompass_Preferred
Molecular Formula C18H19N3O3S, Purity ≥98%, Formula Weight 357.4, Synonyms: BRL 49653; A thazolidinedione agonist of PPAR; binds to the LBD of PPAR (Kd = 43 nM); selectively activates chimeras containing the LBDs of PPAR over PPAR and PPAR in a cell-based reporter assay at 10 M; activates full-length PPAR1 and PPAR2 in a reporter assay (EC50s = 30 and 100 nM, respectively); induces differentiation of C3H10T1/2 stem cells to adipocytes at 1 M; decreases HbA1c and fasting blood glucose levels in a rat model of type 2 diabetes induced by STZ and a high-carbohydrate/high-fat diet at 4 mg/kg; inhibits increases in contusion volume, macrophage infiltration and activation of microglia, and expression of IL-6, MCP1, ICAM1, caspase-3, and Bax in mouse cerebral cortex in a model of traumatic brain injury induced by controlled cortical impact at 6 mg/kg.
Catalog No. NC0950281