Learn More
Medchemexpress LLC HY-17386 200mg , Rosiglitazone BRL49653 CAS:122320-73-4 Purity:98%
SDP
Catalog No. 501492558
Encompass_Preferred
Medchemexpress, HY-17386 200mg Rosiglitazone BRL49653 CAS:122320-73-4 Formula:C18H19N3O3S EC50: 30 nM (PPARγ1), 100 nM (PPARγ2) , 60 nM (PPARγ) , appr 30 nM (TRPC5) Kd: appr 40 nM (PPARγ) IC50: appr 22.5 μM (TRPM2) Purity:98% Rosiglitazone is a potent and selective activator of PPARγ , with EC 50 s of 30 nM, 100 nM and 60 nM for PPARγ1 , PPARγ2 , and PPARγ , respectively, and a K d of appr 40 nM for PPARγ ; Rosiglitazone is also an modulator of TRP channels , inhibits TRP melastatin 2 (TRPM2), TRPM3 and activates TRP canonical 5 (TRPC5). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2558 Supplier Medchemexpress LLC Supplier No. HY17386200MG
Medchemexpress, HY-17386 200mg Rosiglitazone BRL49653 CAS:122320-73-4 Formula:C18H19N3O3S EC50: 30 nM (PPARγ1), 100 nM (PPARγ2) , 60 nM (PPARγ) , appr 30 nM (TRPC5) Kd: appr 40 nM (PPARγ) IC50: appr 22.5 μM (TRPM2) Purity:98% Rosiglitazone is a potent and selective activator of PPARγ , with EC 50 s of 30 nM, 100 nM and 60 nM for PPARγ1 , PPARγ2 , and PPARγ , respectively, and a K d of appr 40 nM for PPARγ ; Rosiglitazone is also an modulator of TRP channels , inhibits TRP melastatin 2 (TRPM2), TRPM3 and activates TRP canonical 5 (TRPC5). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.