Learn More
Apexbio Technology LLC Rosiglitazone HCl 302543-62-0 25mg
SDP
Catalog No. 502240313
Encompass_Preferred
Rosiglitazone HCl (CAS 302543-62-0) is a small-molecule agonist targeting peroxisome proliferator-activated receptor gamma (PPAR ) It is designed to selectively activate PPAR thereby modulating gene transcription involved in glucose metabolism adipogenesis and insulin sensitivity Rosiglitazone HCl exerts its biological activity primarily through activation of PPAR receptors expressed in adipocytes Based on these pharmacological properties Rosiglitazone HCl holds research potential in diabetes mellitus metabolic syndrome obesity and associated metabolic disorders
This item is not returnable. View return policy
Catalog No. 50-224-0313 Supplier Apexbio Technology LLC Supplier No. A431025
Rosiglitazone HCl (CAS 302543-62-0) is a small-molecule agonist targeting peroxisome proliferator-activated receptor gamma (PPAR ) It is designed to selectively activate PPAR thereby modulating gene transcription involved in glucose metabolism adipogenesis and insulin sensitivity Rosiglitazone HCl exerts its biological activity primarily through activation of PPAR receptors expressed in adipocytes Based on these pharmacological properties Rosiglitazone HCl holds research potential in diabetes mellitus metabolic syndrome obesity and associated metabolic disorders