Learn More
Medchemexpress LLC Rosiglitazone hydrochloride | 302543-62-0 | MFCD04112701 | 99.8% | 393.89 | 1 ML
SDP
Catalog No. 5000384578
Encompass_Preferred
Rosiglitazone hydrochloride is an orally active, selective PPARγ agonist. It also acts as a TRPC5 activator and a TRPM3 inhibitor. This compound is used in research for obesity, diabetes, senescence, and ovarian cancer.
  • Selective PPARγ agonist.
  • TRPC5 activator.
  • TRPM3 inhibitor.
  • Used in obesity research.
  • Used in diabetes research.
  • Used in senescence research.
  • Used in ovarian cancer research.
Catalog No. 50-003-84578 Supplier Medchemexpress LLC Supplier No. HY17386A10MM/1ML
Rosiglitazone hydrochloride is an orally active, selective PPARγ agonist. It also acts as a TRPC5 activator and a TRPM3 inhibitor. This compound is used in research for obesity, diabetes, senescence, and ovarian cancer.
  • Selective PPARγ agonist.
  • TRPC5 activator.
  • TRPM3 inhibitor.
  • Used in obesity research.
  • Used in diabetes research.
  • Used in senescence research.
  • Used in ovarian cancer research.