Learn More
Selleck Chemical LLC Rosiglitazone maleate S2505-10mM/1mL
SDP
Catalog No. 503112716
Encompass_Preferred
Rosiglitazone maleate a member of the thiazolidinedione class of antihyperglycaemic agents is a high-affinity selective agonist of the peroxisome proliferator-activated receptor- (PPAR- ) with IC50 of 42 nM Rosiglitazone maleate also modulates TRP channels and induces autophagy Rosiglitazone prevents ferroptosis
Catalog No. 50-311-2716 Supplier Selleck Chemical LLC Supplier No. S250510MM/1ML
Rosiglitazone maleate a member of the thiazolidinedione class of antihyperglycaemic agents is a high-affinity selective agonist of the peroxisome proliferator-activated receptor- (PPAR- ) with IC50 of 42 nM Rosiglitazone maleate also modulates TRP channels and induces autophagy Rosiglitazone prevents ferroptosis