Learn More
Apexbio Technology LLC Rosiglitazone maleate(Synonyms: Avandia, BRL 49653, BRL49653C, Rosiglitazone maleate salt, BRL49653, RGZ, Rosiglizole maleate), 25mg, CAS: 155141-29-0.
SDP
Catalog No. 5000060939
Encompass_Preferred
Rosiglitazone maleate (CAS 155141-29-0) is the maleate salt formulation of rosiglitazone a thiazolidinedione compound targeting the nuclear receptor peroxisome proliferator-activated receptor gamma (PPAR ) By selective activation of PPAR rosiglitazone increases insulin sensitivity of peripheral tissues facilitating glucose uptake and reduction of blood glucose levels Due to its insulin-sensitizing activity rosiglitazone maleate serves as an important research tool in diabetes and metabolic syndrome investigations However recent clinical studies also report associations with adverse cardiovascular events fluid retention and anemia underscoring considerations for risk-benefit evaluation in ongoing research
Catalog No. 50-000-60939 Supplier Apexbio Technology LLC Supplier No. A430225
Rosiglitazone maleate (CAS 155141-29-0) is the maleate salt formulation of rosiglitazone a thiazolidinedione compound targeting the nuclear receptor peroxisome proliferator-activated receptor gamma (PPAR ) By selective activation of PPAR rosiglitazone increases insulin sensitivity of peripheral tissues facilitating glucose uptake and reduction of blood glucose levels Due to its insulin-sensitizing activity rosiglitazone maleate serves as an important research tool in diabetes and metabolic syndrome investigations However recent clinical studies also report associations with adverse cardiovascular events fluid retention and anemia underscoring considerations for risk-benefit evaluation in ongoing research