Learn More
AdipoGen Rosiglitazone maleate
SDP
Catalog No. 502562284
Encompass_Preferred
Chemical. CAS 155141-29-0. Formula C18H19N3O3S . C4H4O4. MW 357.4 . 116.1. Same activities as rosaglitazone Prod. No. AG-CR1-3570 but different formulation. Antidiabetic, hypoglycemic agent. Potent and selective peroxisome proliferator-activated receptor gamma PPAR-gamma agonist. Potent insulin sensitizing agent binding to the PPAR receptors in fat cells and making the cells more responsive to insulin. Ameliorates insulin resistance. Improves blood pressure and vascular function. Enhances proliferation of endogenous neural progenitor cells NPCs. Anti-inflammatory compound. Has controversial therapeutic effects on the cardiovascular system. Promotes adipocyte differentiation of mesenchymal stem cells MSCs.
Catalog No. 50-256-2284 Supplier AdipoGen Supplier No. AGCR13571M100
Chemical. CAS 155141-29-0. Formula C18H19N3O3S . C4H4O4. MW 357.4 . 116.1. Same activities as rosaglitazone Prod. No. AG-CR1-3570 but different formulation. Antidiabetic, hypoglycemic agent. Potent and selective peroxisome proliferator-activated receptor gamma PPAR-gamma agonist. Potent insulin sensitizing agent binding to the PPAR receptors in fat cells and making the cells more responsive to insulin. Ameliorates insulin resistance. Improves blood pressure and vascular function. Enhances proliferation of endogenous neural progenitor cells NPCs. Anti-inflammatory compound. Has controversial therapeutic effects on the cardiovascular system. Promotes adipocyte differentiation of mesenchymal stem cells MSCs.