Learn More
Rosin, Thermo Scientific Chemicals
Description
Rosin is an ingredient in printing inks, photocopying and laser printing paper, varnishes, adhesives (glues), soap, paper sizing, soda, soldering fluxes and sealing wax. It can be used as a glazing agent in medicines and chewing gum. In industry, rosin is a flux used in soldering. A mixture of pitch and rosin is used to make a surface against which glass is polished when making optical components such as lenses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 kg |
Merck Index | 14,8266 |
Solubility Information | Soluble in alcohol, benzene, ether; Insoluble in water |
Physical Form | Powder or Solid |
Chemical Name or Material | Rosin |
Safety and Handling
Hazard Category | H317 |
Hazard Statement | GHS H Statement H317 May cause an allergic skin reaction. |
Precautionary Statement | P261-P272-P280g-P302+P352-P333+P313-P363-P501c |
EINECSNumber | 232-475-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.