Learn More
Electron Microscopy Sciences Rosin Alcohol Stock Solution 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2613202
Encompass_Preferred
Used in Wolbach's Giemsa Method Wolbach, Todd, and Palfrey (1922) Nuclei, collagen. References are Wolbach, S.B., Todd, J.L. and Palfrey, F.W., The Etilogy of Pathology Typhus, Harvard University Press, Cambridge, MA., p. 13 – 14,c 1922 and Luna, L.G., (ed), Manual of Histologic Staining Methods of the AFIP, 3rd Edition, McGraw-Hill, Y, p. 119, c. 1968
Catalog No. 50-316-89