Learn More
RS AMERICAS INC ROSIN CORE SOLDER WIRE; SN60PB
Catalog No. NC1742720
Encompass
Rosin Core Solder Wire; Sn60Pb40 Alloy; 60/40; OD 0.062"(1.57mm); Melt at 650-750F
Catalog No. NC1742720 Supplier RS AMERICAS INC Supplier No. 70054236
Rosin Core Solder Wire; Sn60Pb40 Alloy; 60/40; OD 0.062"(1.57mm); Melt at 650-750F