Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rosin, Gum, MP Biomedicals™

Catalog No. ICN15655790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15655790 500 g
ICN15655791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15655790 Supplier MP Biomedicals, Inc Supplier No. 0215655790
Add to Cart
Add to Cart

acid value: 150 to 170

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8050-09-7
Molecular Formula C20H30O2
Molecular Weight (g/mol) 302.46
MDL Number MFCD00132205
InChI Key RSWGJHLUYNHPMX-UHFFFAOYNA-N
Synonym 4bs,1r,10ar,4ar-1,4a-dimethyl-7-methylethyl-1,2,3,4,5,6,10,10a,4a,4b-decah ydrophenanthrenecarboxylic acid, 4ar-7-isopropyl-1,4a-dimethyl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid

Specifications

Color Orange
Density 1.07 - 1.09g/cm3 at 20°C
Quantity 500 g
Physical Form Chunks
Chemical Name or Material Rosin, Gum
EINECSNumber 232-475-7
Recommended Storage Store at Room Temperature (15 to 30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.