Learn More
TARGETMOL CHEMICALS INC ROSLIN 2 BROMIDE 5MG
SDP
Catalog No. 502580787
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Roslin 2 bromide (Benzylhexamethylenetetramine bromide) is a p53 reactivator that disrupts the binding of FAK and p5. Roslin 2 bromide exhibits anticancer effects. purity: 97%
Catalog No. 50-258-0787 Supplier TARGETMOL CHEMICALS INC Supplier No. T247305MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Roslin 2 bromide (Benzylhexamethylenetetramine bromide) is a p53 reactivator that disrupts the binding of FAK and p5. Roslin 2 bromide exhibits anticancer effects. purity: 97%