Learn More
Medchemexpress LLC HY-N5015 5mg Medchemexpress, Rosmanol CAS:80225-53-2 Purity:>98%
SDP
Catalog No. 501933954
Encompass_Preferred
Medchemexpress, HY-N5015 5mg Rosmanol CAS:80225-53-2 Rosmanol could inhibit the oxidation of low density lipoprotein (LPL) and significantly inhibit lipopolysaccharide induced iNOS and COX-2 expression, with anti-inflammatory effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3954 Supplier Medchemexpress LLC Supplier No. HYN50155MG
Medchemexpress, HY-N5015 5mg Rosmanol CAS:80225-53-2 Rosmanol could inhibit the oxidation of low density lipoprotein (LPL) and significantly inhibit lipopolysaccharide induced iNOS and COX-2 expression, with anti-inflammatory effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.