Learn More
Cayman Chemical RosmarInIc AcId 10mg
SDP
Catalog No. 502896919
Encompass_Preferred
Rosmarinic acid is a naturally-occurring phenolic compound with antioxidant and anti-inflammatory properties. This compound inhibits lipid peroxidation of rat liver microsomes by 90% at a concentration of 25 UG/ml.{4778} Rosmarinic acid suppresses endotoxin-induced activation of complement and concomitant formation of prostacyclin.{4407,4408} Formation of 5-HETE and LTB4 from human PMNL is inhibited by rosmarinic acid at concentrations of 10-5 to 10-3 M.{4706}
Catalog No. 50-289-6919 Supplier Cayman Chemical Supplier No. 7090010 MG
Rosmarinic acid is a naturally-occurring phenolic compound with antioxidant and anti-inflammatory properties. This compound inhibits lipid peroxidation of rat liver microsomes by 90% at a concentration of 25 UG/ml.{4778} Rosmarinic acid suppresses endotoxin-induced activation of complement and concomitant formation of prostacyclin.{4407,4408} Formation of 5-HETE and LTB4 from human PMNL is inhibited by rosmarinic acid at concentrations of 10-5 to 10-3 M.{4706}